Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wbscr27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024479.2 | NP_077799.2 | 3 - 4 | 389 | 105 | |
GenBank mRNA | AK042578.1 | - | 3 - 4 | 280 | 105 | |
AK081683.1 | - | 3 - 4 | 408 | 105 | ||
AK132481.1 | - | 3 - 4 | 378 | 105 | ||
AY138964.1 | - | 3 - 4 | 389 | 105 | ||
AY354925.2 | - | 3 - 4 | 389 | 105 | ||
AY354926.1 | - | 3 - 4 | 290 | 105 | ||
AY354927.1 | - | 3 - 4 | 389 | 105 | ||
BC002286.1 | - | 3 - 4 | 380 | 3589835 | 105 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |