Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Blvra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026678.4 | NP_080954.4 | 3 - 4 | 325 | 102 | |
GenBank mRNA | AK002231.1 | - | 2 - 3 | 264 | 102 | |
AK010847.1 | - | 3 - 4 | 280 | 102 | ||
AK045022.1 | - | 3 - 4 | 318 | 102 | ||
AK162186.1 | - | 3 - 4 | 286 | 102 | ||
AK172620.1 | - | 3 - 4 | 290 | 102 | ||
BC052146.1 | - | 3 - 4 | 315 | 6588485 | 102 | |
CA983696.1 | - | 1 - 2 | 60 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |