Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpne2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153507.2 | NP_705727.1 | 3 - 4 | 566 | 70 | |
GenBank mRNA | BC022128.1 | - | 3 - 4 | 527 | 5097527 | 70 |
BC031801.1 | - | 3 - 4 | 566 | 3985959 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |