Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gdpd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024228.2 | NP_077190.2 | 6 - 7 | 631 | 64 | |
GenBank mRNA | AK003726.1 | - | 6 - 7 | 633 | 64 | |
BC002172.1 | - | 4 - 5 | 550 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |