Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phf10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024250.4 | NP_077212.3 | 6 - 7 | 762 | 80 | |
GenBank mRNA | AB221576.1 | - | 4 - 5 | 429 | 80 | |
AK007873.1 | - | 6 - 7 | 742 | 80 | ||
BC002206.1 | - | 4 - 5 | 496 | 3489870 | 80 | |
CA322732.1 | - | 3 - 4 | 320 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |