Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026167.4 | NP_080443.1 | 3 - 4 | 290 | 64 | |
GenBank mRNA | AK004677.1 | - | 3 - 4 | 262 | 64 | |
BY136647.1 | - | 3 - 4 | 290 | 64 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus Inventoried Probe spans exons |