Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rer1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026395.1 | NP_080671.1 | 2 - 3 | 182 | 89 | |
GenBank mRNA | AK002550.1 | - | 2 - 3 | 182 | 89 | |
AK003682.1 | - | 2 - 3 | 204 | 89 | ||
AK013262.1 | - | 2 - 3 | 239 | 89 | ||
AK078396.1 | - | 2 - 3 | 245 | 89 | ||
AK168192.1 | - | 3 - 4 | 401 | 89 | ||
BC029189.1 | - | 1 - 2 | 88 | 4036721 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |