Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zcchc10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026479.4 | NP_080755.2 | 2 - 3 | 215 | 117 | |
GenBank mRNA | AK010803.1 | - | 2 - 3 | 226 | 117 | |
AK020177.1 | - | 2 - 3 | 216 | 117 | ||
AK027985.1 | - | 2 - 3 | 223 | 117 | ||
AK169856.1 | - | 2 - 3 | 216 | 117 | ||
BC025078.1 | - | 2 - 3 | 231 | 4504126 | 117 | |
BC087902.1 | - | 2 - 3 | 215 | 30505854 | 117 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |