Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0200026425.mm.1 |
NM_001037098.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200026425.mm.1 |
NM_001037098.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200026425.mm.1 |
NM_026495.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200026425.mm.1 |
NM_026495.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200026425.mm.1 |
TC0200003176.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200026432.mm.1 |
NM_026495.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200026432.mm.1 |
NM_026495.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200026432.mm.1 |
TC0200003176.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200026422.mm.1 |
NM_001037098.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200026422.mm.1 |
NM_001037098.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200026422.mm.1 |
NM_026495.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200026422.mm.1 |
NM_026495.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200026422.mm.1 |
TC0200003176.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200026423.mm.1 |
NM_001037098.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200026423.mm.1 |
NM_001037098.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200026423.mm.1 |
NM_026495.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200026423.mm.1 |
NM_026495.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200026423.mm.1 |
TC0200003176.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200026426.mm.1 |
NM_001037098.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200026426.mm.1 |
NM_001037098.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200026426.mm.1 |
NM_026495.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200026426.mm.1 |
NM_026495.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200026426.mm.1 |
TC0200003176.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200026427.mm.1 |
NM_001037098.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200026427.mm.1 |
NM_001037098.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200026427.mm.1 |
NM_026495.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200026427.mm.1 |
NM_026495.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200026427.mm.1 |
TC0200003176.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200026424.mm.1 |
NM_001037098.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200026424.mm.1 |
NM_001037098.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200026424.mm.1 |
NM_026495.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200026424.mm.1 |
NM_026495.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200026424.mm.1 |
TC0200003176.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0200026421.mm.1 |
NM_001037098.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200026421.mm.1 |
NM_001037098.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200026421.mm.1 |
NM_026495.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200026421.mm.1 |
NM_026495.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0200026421.mm.1 |
TC0200003176.mm.1 |
MTA TC |