Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026542.3 | NP_080818.1 | 7 - 8 | 666 | 81 | |
GenBank mRNA | AK007934.1 | - | 7 - 8 | 620 | 81 | |
AK013834.1 | - | 3 - 4 | 310 | 81 | ||
AK019396.1 | - | 7 - 8 | 658 | 81 | ||
AK078417.1 | - | 7 - 8 | 658 | 81 | ||
AK084116.1 | - | 7 - 8 | 657 | 81 | ||
AK160922.1 | - | 7 - 8 | 611 | 81 | ||
AK161289.1 | - | 7 - 8 | 658 | 81 | ||
AK211485.1 | - | 2 - 3 | 149 | 81 | ||
BC115661.1 | - | 7 - 8 | 566 | 40103267 | 81 | |
BC115662.1 | - | 7 - 8 | 573 | 40103268 | 81 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |