Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnaaf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026648.4 | NP_080924.1 | 9 - 10 | 1477 | 68 | |
GenBank mRNA | AK006656.1 | - | 2 - 3 | 136 | 68 | |
AK019633.1 | - | 9 - 10 | 1477 | 68 | ||
BC050751.1 | - | 9 - 10 | 1396 | 6774345 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |