Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cep89 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028120.2 | NP_082396.1 | 13 - 14 | 1457 | 84 | |
GenBank mRNA | AK012067.1 | - | 13 - 14 | 1458 | 84 | |
AK029107.1 | - | 13 - 14 | 1488 | 84 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons |