Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc19a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030556.2 | NP_085033.2 | 3 - 4 | 361 | 82 | |
GenBank mRNA | AF271634.1 | - | 3 - 4 | 361 | 82 | |
AK098112.1 | - | 3 - 4 | 356 | 82 | ||
BC109154.1 | - | 3 - 4 | 344 | 40047017 | 82 | |
BC109155.1 | - | 2 - 3 | 214 | 40047019 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |