Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rad54l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030730.2 | NP_109655.2 | 16 - 17 | 2939 | 72 | |
GenBank mRNA | AJ132389.1 | - | 16 - 17 | 2840 | 72 | |
AK083903.1 | - | 16 - 17 | 2921 | 72 | ||
AK143506.1 | - | 15 - 16 | 2555 | 72 | ||
AK155151.1 | - | 16 - 17 | 2896 | 72 | ||
BC133714.1 | - | 16 - 17 | 2788 | 40143565 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |