Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mmp1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_032006.3 | NP_114395.1 | 3 - 4 | 543 | 101 | |
GenBank mRNA | AJ278462.1 | - | 3 - 4 | 497 | 101 | |
AK049552.1 | - | 3 - 4 | 544 | 101 | ||
BC117756.2 | - | 3 - 4 | 543 | 40090601 | 101 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |