Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: M1ap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033079.2 | NP_149070.2 | 4 - 5 | 679 | 67 | |
GenBank mRNA | AF084364.1 | - | 3 - 4 | 648 | 67 | |
AK131954.1 | - | 4 - 5 | 679 | 67 | ||
BC119803.1 | - | 3 - 4 | 606 | 40107692 | 67 | |
BC122521.1 | - | 3 - 4 | 611 | 40107688 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |