Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prcc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_033573.2 | NP_291051.1 | 5 - 6 | 1546 | 78 | |
GenBank mRNA | AF273750.1 | - | 5 - 6 | 1567 | 78 | |
AK078394.1 | - | 5 - 6 | 1566 | 78 | ||
BC052072.1 | - | 5 - 6 | 1546 | 5717482 | 78 | |
BC082784.1 | - | 5 - 6 | 1564 | 4191155 | 78 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |