Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fgd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053072.3 | NP_444302.4 | 18 - 19 | 4172 | 70 | |
GenBank mRNA | AK016940.1 | - | 12 - 13 | 1145 | 70 | |
AK173157.1 | - | 18 - 19 | 4012 | 70 | ||
BC026860.1 | - | 17 - 18 | 2510 | 70 | ||
BC139019.1 | - | 18 - 19 | 3966 | 8862041 | 70 | |
BC139022.1 | - | 18 - 19 | 3966 | 8862044 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |