Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Entpd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053103.5 | NP_444333.3 | 11 - 12 | 1726 | 93 | |
GenBank mRNA | AK081564.1 | - | 11 - 12 | 1584 | 93 | |
AK147401.1 | - | 11 - 12 | 1591 | 93 | ||
AK170546.1 | - | 11 - 12 | 1744 | 93 | ||
BC137709.1 | - | 10 - 11 | 1512 | 8860728 | 93 | |
BC137717.1 | - | 10 - 11 | 1512 | 8860736 | 93 |
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
---|---|---|---|---|---|---|---|
GenBankmRNA | AF288221.1 | - | 1630 | Chr. - - [-] on Build | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |