Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tle6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053254.2 | NP_444484.1 | 12 - 13 | 1704 | 73 | |
GenBank mRNA | AB221587.1 | - | 11 - 12 | 1565 | 73 | |
AF145957.2 | - | 12 - 13 | 1704 | 73 | ||
AK007880.1 | - | 6 - 7 | 936 | 73 | ||
AK154391.1 | - | 9 - 10 | 1665 | 73 | ||
BC023315.1 | - | 12 - 13 | 1603 | 73 | ||
BC048832.1 | - | 9 - 10 | 1543 | 4506029 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |