Array Name |
Probe IDs |
Targets |
Target Type |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27994101 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27994102 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27994097 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27994099 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27994103 |
- |
- |
Agilent Whole Mouse Genome Microarray 4x44K v2 |
A_55_P1955891 |
BC090648.1 |
mouse_transcriptome |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
886066 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
887782 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
236422 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
131258 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
496916 |
D10584.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
587939 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
233568 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
948326 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
376192 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
140792 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
654243 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
56562 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
444211 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
316151 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
214433 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
164017 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
875808 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
931277 |
AK004392.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
931277 |
AK041053.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
931277 |
AK081513.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
931277 |
AK156350.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
931277 |
AK167557.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
931277 |
BC005536.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
931277 |
BC019131.1 |
genbank_mrna_mouse |
Agilent SurePrint G3 Mouse CGH Microarray 1x1M |
931277 |
NM_173752.4 |
refseq_mrna_mouse_NM |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
26213 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
174382 |
- |
- |
Agilent SurePrint G3 Mouse CGH Microarray 4x180K |
21139 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P144601 |
- |
- |
Agilent Mouse Genome CGH Microarray 44K G4426B |
A_53_P141198 |
- |
- |
SurePrint G3 Mouse Gene Expression Microarray G4852A |
A_55_P1955891 |
BC090648.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P252519 |
BC141034.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P252519 |
BC090648.1 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P252519 |
NM_053269.3 |
mouse_transcriptome |
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " |
A_51_P252519 |
AK076551.1 |
mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27994096 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27994113 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27994106 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27994094 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27994108 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27994083 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27994111 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27994086 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27994088 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27994098 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27994104 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27994120 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27994112 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27994109 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27994099 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27994092 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27994101 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27994116 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27994114 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27994103 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27994117 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27994095 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27994085 |
- |
- |
Agilent Mouse Promoter ChIP-on-Chip A) |
A_68_P27994090 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P144601 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P07431069 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P07431041 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_67_P07431057 |
- |
- |
Agilent Mouse Genome CGH Microarray 244A (G4415A) |
A_53_P141198 |
- |
- |
Whole Mouse Genome G4122A |
A_51_P252519 |
BC141034.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P252519 |
BC090648.1 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P252519 |
NM_053269.3 |
mouse_transcriptome |
Whole Mouse Genome G4122A |
A_51_P252519 |
AK076551.1 |
mouse_transcriptome |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P120796 |
- |
- |
Agilent Mouse Developmental Microarray 44K G2519A Option 002 |
A_66_P110814 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P141198 |
- |
- |
Agilent Mouse Genome CGH Microarray 105A (G4416A) |
A_53_P144601 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P09120053 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P09648111 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P08974795 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P09214880 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P09214881 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P09648113 |
- |
- |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) |
A_68_P09120052 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27994099 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27994101 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27994102 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27994097 |
- |
- |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K |
A_68_P27994103 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27994071 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27994028 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27994036 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27994052 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27993954 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27993939 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27994094 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27994008 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27993987 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27994082 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27993969 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27994009 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27993977 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27994038 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27994007 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27994064 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27994095 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27994098 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27994083 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27994002 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27994022 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27994011 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27994004 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27993986 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27993955 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27994073 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27993968 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27994029 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27994021 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27993973 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27993946 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27994099 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27994069 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27994045 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27994000 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27994088 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27994037 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27993958 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27994091 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27994020 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27993957 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27994101 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27994032 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27994001 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27994085 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27993965 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27994057 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27993950 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27994044 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27993942 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27993949 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27994081 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27993971 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27994104 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27993966 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27994065 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27994041 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27994017 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27993980 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27993941 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27994103 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27993982 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27994027 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27993990 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27994031 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27993947 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27994061 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27994026 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27993984 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27994024 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27994092 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27993943 |
- |
- |
Agilent Mouse Whole Genome ChIP-on-Chip |
A_68_P27994035 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27994013 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27994073 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27994029 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27994027 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27993960 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27994094 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27994095 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27994056 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27993957 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27993951 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09630072 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27994088 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27994046 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27993956 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27994018 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27994101 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27994063 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09410012 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27994019 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09024124 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27993971 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09262341 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09345489 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27994085 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27993969 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27994028 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27994035 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27994081 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27993966 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27994082 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27993954 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27994031 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27994044 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27994038 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27994061 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27993961 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27993955 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27994076 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27994022 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27994055 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27994041 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27994032 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27994014 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09645404 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09714839 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27993965 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27994020 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09458932 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27994075 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27994092 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27994103 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27994025 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27994021 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27994016 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27994069 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27994015 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27994098 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27993973 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27994086 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27994077 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27994024 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27994026 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27994099 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27994045 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P27993968 |
- |
- |
Agilent Six_ChIP_02_of_03_v2 |
A_68_P09357381 |
- |
- |