Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Foxp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001199347.1 | NP_001186276.1 | 10 - 11 | 1158 | 80 | |
NM_001199348.1 | NP_001186277.1 | 9 - 10 | 1016 | 80 | ||
NM_054039.2 | NP_473380.1 | 9 - 10 | 1091 | 80 | ||
GenBank mRNA | AF277991.1 | - | 9 - 10 | 1016 | 80 | |
AF277992.1 | - | 9 - 10 | 1074 | 80 | ||
AK157447.1 | - | 10 - 11 | 1159 | 80 | ||
AY357712.1 | - | 8 - 9 | 874 | 80 | ||
AY357713.1 | - | 8 - 9 | 874 | 80 | ||
BC132333.1 | - | 8 - 9 | 862 | 40130610 | 80 | |
BC132335.1 | - | 8 - 9 | 862 | 40130612 | 80 | |
DQ387959.1 | - | 10 - 11 | 1158 | 80 | ||
KF471319.1 | - | 8 - 9 | 858 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons |