Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc13a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_054055.2 | NP_473396.1 | 4 - 5 | 648 | 77 | |
GenBank mRNA | AF306491.1 | - | 4 - 5 | 647 | 77 | |
AK085326.1 | - | 4 - 5 | 646 | 77 | ||
AK087518.1 | - | 4 - 5 | 646 | 77 | ||
AK128982.1 | - | 4 - 5 | 645 | 77 | ||
AK138344.1 | - | 4 - 5 | 646 | 77 | ||
AK157881.1 | - | 4 - 5 | 617 | 77 | ||
AK165355.1 | - | 4 - 5 | 646 | 77 | ||
BC026803.1 | - | 4 - 5 | 634 | 4238769 | 77 | |
BC028882.1 | - | 4 - 5 | 632 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |