Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc13a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_054055.2 | NP_473396.1 | 5 - 6 | 823 | 114 | |
GenBank mRNA | AF306491.1 | - | 5 - 6 | 822 | 114 | |
AK085326.1 | - | 5 - 6 | 821 | 114 | ||
AK087518.1 | - | 5 - 6 | 821 | 114 | ||
AK128982.1 | - | 5 - 6 | 820 | 114 | ||
AK138344.1 | - | 5 - 6 | 821 | 114 | ||
AK157881.1 | - | 5 - 6 | 792 | 114 | ||
BC026803.1 | - | 5 - 6 | 809 | 4238769 | 114 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |