Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elmo2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_080287.2 | NP_525026.2 | 15 - 16 | 1510 | 77 | |
NM_207706.1 | NP_997589.1 | 16 - 17 | 1546 | 77 | ||
GenBank mRNA | AB093301.1 | - | 6 - 7 | 2091 | 77 | |
AF398884.1 | - | 14 - 15 | 1409 | 77 | ||
AK032033.1 | - | 16 - 17 | 1519 | 77 | ||
AK033918.1 | - | 15 - 16 | 1704 | 77 | ||
AK038455.1 | - | 15 - 16 | 1616 | 77 | ||
AK045428.1 | - | 15 - 16 | 1511 | 77 | ||
AK047040.1 | - | 15 - 16 | 1472 | 77 | ||
AK050823.1 | - | 15 - 16 | 1473 | 77 | ||
AK053574.1 | - | 16 - 17 | 1510 | 77 | ||
AK129023.1 | - | 15 - 16 | 1510 | 77 | ||
AK200380.1 | - | 2 - 3 | 192 | 77 | ||
BC023954.1 | - | 16 - 17 | 1531 | 5320429 | 77 | |
BC033418.1 | - | 14 - 15 | 1440 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |