Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adcy6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007405.2 | NP_031431.2 | 15 - 16 | 2589 | 83 | |
GenBank mRNA | AK147516.1 | - | 15 - 16 | 2589 | 83 | |
M93422.1 | - | 14 - 15 | 2521 | 83 | ||
M96653.1 | - | 14 - 15 | 2411 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Mus musculus Probe spans exons |