Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nxn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008750.5 | NP_032776.1 | 4 - 5 | 792 | 79 | |
GenBank mRNA | AB095441.1 | - | 4 - 5 | 637 | 79 | |
AB095442.1 | - | 4 - 5 | 637 | 79 | ||
AB095443.1 | - | 4 - 5 | 637 | 79 | ||
AB095444.1 | - | 4 - 5 | 637 | 79 | ||
AK012352.1 | - | 4 - 5 | 777 | 79 | ||
AK149075.1 | - | 4 - 5 | 776 | 79 | ||
BC004688.1 | - | 4 - 5 | 752 | 3501314 | 79 | |
BC047339.1 | - | 3 - 4 | 324 | 79 | ||
BC058244.1 | - | 4 - 5 | 792 | 6312910 | 79 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |