Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ap3s2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009682.3 | NP_033812.3 | 1 - 2 | 135 | 73 | |
GenBank mRNA | AK028372.1 | - | 1 - 2 | 118 | 73 | |
AK037336.1 | - | 1 - 2 | 113 | 73 | ||
AK083911.1 | - | 1 - 2 | 124 | 73 | ||
AK150085.1 | - | 1 - 2 | 128 | 73 | ||
BC060236.1 | - | 1 - 2 | 96 | 6853219 | 73 | |
U91933.1 | - | 1 - 2 | 90 | 73 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |