Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nell2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289653.1 | NP_001276582.1 | 5 - 6 | 560 | 75 | |
GenBank mRNA | AK035019.1 | - | 3 - 4 | 413 | 75 | |
AK039790.1 | - | 624 | 75 | |||
AK048913.1 | - | 831 | 75 | |||
BC051968.1 | - | 4 - 5 | 528 | 6401465 | 75 | |
U59230.1 | - | 5 - 6 | 561 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |