Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srcin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_018873.2 | NP_061361.2 | 14 - 15 | 2716 | 145 | |
GenBank mRNA | AF040944.1 | - | 14 - 15 | 2718 | 145 | |
AK129422.1 | - | 9 - 10 | 1873 | 145 | ||
U59873.1 | - | 14 - 15 | 2717 | 145 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Detect Genomic DNA Mus musculus Probe spans exons |