Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Btd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025295.4 | NP_079571.1 | 1 - 2 | 73 | 90 | |
GenBank mRNA | AK005506.1 | - | 1 - 2 | 66 | 90 | |
AK135375.1 | - | 1 - 2 | 63 | 90 | ||
CJ181966.1 | - | 1 - 2 | 73 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mus musculus Inventoried Probe spans exons |