Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eri1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026067.3 | NP_080343.4 | 1 - 2 | 266 | 80 | |
GenBank mRNA | AK014041.1 | - | 1 - 2 | 267 | 80 | |
AK014410.1 | - | 1 - 2 | 268 | 80 | ||
AK144223.1 | - | 1 - 2 | 255 | 80 | ||
AK165026.1 | - | 1 - 2 | 195 | 80 | ||
AK172005.1 | - | 1 - 2 | 233 | 80 | ||
BC046412.1 | - | 1 - 2 | 214 | 5354985 | 80 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |