Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdh16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252627.1 | NP_001239556.1 | 13 - 14 | 2099 | 70 | |
NM_001252628.1 | NP_001239557.1 | 14 - 15 | 2189 | 70 | ||
NM_007663.3 | NP_031689.1 | 14 - 15 | 2189 | 70 | ||
GenBank mRNA | AF016271.1 | - | 14 - 15 | 2057 | 70 | |
AJ609635.1 | - | 14 - 15 | 2056 | 70 | ||
AK052636.1 | - | 14 - 15 | 1992 | 70 | ||
AK093905.1 | - | 14 - 15 | 1987 | 70 | ||
AK164546.1 | - | 13 - 14 | 2100 | 70 | ||
BC015251.1 | - | 14 - 15 | 2037 | 4235757 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |