Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cenpa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AA016357.1 | - | 2 - 3 | 263 | 75 | |
AF012709.1 | - | 2 - 3 | 319 | 75 | ||
AK010718.1 | - | 2 - 3 | 281 | 75 | ||
AK011399.1 | - | 2 - 3 | 264 | 75 | ||
AK040313.1 | - | 3 - 4 | 446 | 75 | ||
AK041138.1 | - | 3 - 4 | 439 | 75 | ||
AK053763.1 | - | 2 - 3 | 288 | 75 | ||
AK089033.1 | - | 2 - 3 | 292 | 75 | ||
BC011038.1 | - | 2 - 3 | 299 | 4018429 | 75 | |
BC012280.1 | - | 4 - 5 | 392 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Mus musculus Probe spans exons |