Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbln2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081437.1 | NP_001074906.1 | 13 - 14 | 2949 | 56 | |
NM_007992.2 | NP_032018.2 | 14 - 15 | 3090 | 56 | ||
GenBank mRNA | AK087922.1 | - | 2 - 3 | 335 | 56 | |
AK159361.1 | - | 13 - 14 | 3261 | 56 | ||
AK168688.1 | - | 13 - 14 | 2887 | 56 | ||
BC005443.1 | - | 13 - 14 | 2929 | 3490759 | 56 | |
X75285.1 | - | 14 - 15 | 2990 | 56 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |