Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fer | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008000.2 | NP_032026.2 | 5 - 6 | 748 | 148 | |
GenBank mRNA | AK082799.1 | - | 7 - 8 | 959 | 148 | |
AK157828.1 | - | 10 - 11 | 1430 | 148 | ||
M32054.1 | - | 5 - 6 | 748 | 148 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Probe spans exons |