Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sh3pxd2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164717.1 | NP_001158189.1 | 11 - 12 | 1450 | 94 | |
NM_008018.4 | NP_032044.2 | 12 - 13 | 1534 | 94 | ||
GenBank mRNA | AJ007012.1 | - | 12 - 13 | 1534 | 94 | |
AK085227.1 | - | 5 - 6 | 1373 | 94 | ||
BC118022.1 | - | 11 - 12 | 1286 | 40097448 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |