Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gad2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008078.2 | NP_032104.2 | 6 - 7 | 1064 | 66 | |
GenBank mRNA | AF326549.1 | - | 6 - 7 | 728 | 66 | |
AF326550.1 | - | 6 - 7 | 728 | 66 | ||
AK136834.1 | - | 6 - 7 | 764 | 66 | ||
BC018380.1 | - | 6 - 7 | 779 | 4482097 | 66 | |
D42051.1 | - | 6 - 7 | 1080 | 66 | ||
KC134211.1 | - | 6 - 7 | 728 | 66 | ||
L16980.1 | - | 6 - 7 | 800 | 66 | ||
S67454.1 | - | 2 - 3 | 206 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |