Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008636.4 | NP_032662.3 | 6 - 7 | 1133 | 63 | |
GenBank mRNA | AK030284.1 | - | 6 - 7 | 1112 | 63 | |
AK150295.1 | - | 7 - 8 | 1195 | 63 | ||
AY260849.1 | - | 5 - 6 | 987 | 63 | ||
AY260850.1 | - | 5 - 6 | 987 | 63 | ||
BC017679.1 | - | 6 - 7 | 1095 | 4210829 | 63 | |
X71327.1 | - | 6 - 7 | 1083 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |