Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sptbn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009260.2 | NP_033286.2 | 20 - 21 | 4885 | 87 | |
NM_175836.2 | NP_787030.2 | 20 - 21 | 4545 | 87 | ||
GenBank mRNA | AF017112.1 | - | 20 - 21 | 4792 | 87 | |
AK220456.1 | - | 21 - 22 | 4747 | 87 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Mus musculus Probe spans exons |