Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sva | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009299.2 | NP_033325.2 | 2 - 3 | 264 | 110 | |
GenBank mRNA | BC104304.1 | - | 2 - 3 | 262 | 40043712 | 110 |
BC104305.1 | - | 2 - 3 | 264 | 40043713 | 110 | |
CJ055470.1 | - | 2 - 3 | 264 | 110 | ||
M94179.1 | - | 2 - 3 | 250 | 110 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |