Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Capn7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009796.2 | NP_033926.1 | 17 - 18 | 2171 | 119 | |
GenBank mRNA | AB028640.1 | - | 17 - 18 | 2171 | 119 | |
AJ012475.1 | - | 17 - 18 | 1984 | 119 | ||
AK154277.1 | - | 17 - 18 | 2257 | 119 | ||
AK169334.1 | - | 17 - 18 | 3216 | 119 | ||
BC095980.1 | - | 17 - 18 | 3186 | 119 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protease Mammalian Gene Collection Mus musculus Probe spans exons |