Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp46a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010010.1 | NP_034140.1 | 13 - 14 | 1334 | 94 | |
GenBank mRNA | AF094479.1 | - | 13 - 14 | 1334 | 94 | |
AK140740.1 | - | 13 - 14 | 1339 | 94 | ||
BC018307.1 | - | 13 - 14 | 1357 | 4195579 | 94 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |