Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fah | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010176.4 | NP_034306.2 | 3 - 4 | 540 | 70 | |
GenBank mRNA | AK143759.1 | - | 3 - 4 | 369 | 70 | |
AK158808.1 | - | 3 - 4 | 426 | 70 | ||
BC010767.1 | - | 3 - 4 | 325 | 4242421 | 70 | |
M84145.1 | - | 3 - 4 | 366 | 70 | ||
Z11774.1 | - | 3 - 4 | 352 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |