Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fkbp8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111066.1 | NP_001104536.1 | 6 - 7 | 1032 | 68 | |
NM_001199631.1 | NP_001186560.1 | 6 - 7 | 1128 | 68 | ||
NM_010223.2 | NP_034353.2 | 6 - 7 | 1029 | 68 | ||
GenBank mRNA | AB154247.1 | - | 5 - 6 | 768 | 68 | |
AF030635.1 | - | 5 - 6 | 799 | 68 | ||
AK088739.1 | - | 6 - 7 | 967 | 68 | ||
AK146122.1 | - | 6 - 7 | 1027 | 68 | ||
AK152198.1 | - | 6 - 7 | 1028 | 68 | ||
AK153000.1 | - | 6 - 7 | 1025 | 68 | ||
AK165281.1 | - | 6 - 7 | 1025 | 68 | ||
AK167663.1 | - | 6 - 7 | 1015 | 68 | ||
AY225340.1 | - | 5 - 6 | 912 | 68 | ||
AY278608.1 | - | 6 - 7 | 988 | 68 | ||
BC003739.1 | - | 5 - 6 | 948 | 3601511 | 68 | |
BC027808.1 | - | 6 - 7 | 982 | 5324380 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Apoptosis Mammalian Gene Collection Mus musculus Probe spans exons |