Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mag | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010758.2 | NP_034888.1 | 2 - 3 | 118 | 77 | |
GenBank mRNA | AK003029.1 | - | 3 - 4 | 152 | 77 | |
AK045564.1 | - | 2 - 3 | 107 | 77 | ||
BC101944.1 | - | 2 - 3 | 91 | 40039200 | 77 | |
BC103521.1 | - | 2 - 3 | 91 | 40039204 | 77 | |
BC103522.1 | - | 2 - 3 | 91 | 77 | ||
BC103523.1 | - | 2 - 3 | 91 | 77 | ||
M31811.1 | - | 2 - 3 | 118 | 77 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |