Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0900005029.mm.1 |
NM_001110350.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900005029.mm.1 |
NM_001110350.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900005029.mm.1 |
NM_001110351.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900005029.mm.1 |
NM_001110351.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900005029.mm.1 |
NM_011378.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900005029.mm.1 |
NM_011378.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900005029.mm.1 |
TC0900000695.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900005026.mm.1 |
NM_001110351.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900005026.mm.1 |
NM_001110351.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900005026.mm.1 |
NM_011378.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900005026.mm.1 |
NM_011378.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900005026.mm.1 |
TC0900000695.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900005028.mm.1 |
NM_001110350.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900005028.mm.1 |
NM_001110350.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900005028.mm.1 |
NM_001110351.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900005028.mm.1 |
NM_001110351.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900005028.mm.1 |
NM_011378.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900005028.mm.1 |
NM_011378.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900005028.mm.1 |
TC0900000695.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900005027.mm.1 |
NM_001110351.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900005027.mm.1 |
NM_001110351.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900005027.mm.1 |
NM_011378.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900005027.mm.1 |
NM_011378.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900005027.mm.1 |
TC0900000695.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900005023.mm.1 |
NM_001110350.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900005023.mm.1 |
NM_001110350.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900005023.mm.1 |
NM_011378.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900005023.mm.1 |
NM_011378.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900005023.mm.1 |
TC0900000695.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900005024.mm.1 |
NM_001110350.1.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900005024.mm.1 |
NM_001110350.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900005024.mm.1 |
NM_011378.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900005024.mm.1 |
NM_011378.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900005024.mm.1 |
TC0900000695.mm.1 |
MTA TC |