Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Galnt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001160404.1 | NP_001153876.1 | 6 - 7 | 934 | 83 | |
NM_013814.3 | NP_038842.3 | 6 - 7 | 1022 | 83 | ||
GenBank mRNA | AK083391.1 | - | 6 - 7 | 937 | 83 | |
BC056215.1 | - | 6 - 7 | 939 | 30019829 | 83 | |
BC090962.1 | - | 5 - 6 | 812 | 30626890 | 83 | |
U73820.1 | - | 4 - 5 | 687 | 83 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |