Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fut8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001252614.1 | NP_001239543.1 | 5 - 6 | 802 | 71 | |
NM_016893.5 | NP_058589.2 | 4 - 5 | 713 | 71 | ||
NR_045554.1 | 4 - 5 | 1132 | 71 | |||
GenBank mRNA | AB025198.1 | - | 1 - 2 | 402 | 71 | |
AK048520.1 | - | 4 - 5 | 851 | 71 | ||
AK051811.1 | - | 4 - 5 | 670 | 71 | ||
AK134537.1 | - | 4 - 5 | 806 | 71 | ||
AK163494.1 | - | 4 - 5 | 829 | 71 | ||
BC010666.1 | - | 5 - 6 | 781 | 3594582 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons |