Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctps2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168568.1 | NP_001162040.1 | 6 - 7 | 867 | 75 | |
NM_001168569.1 | NP_001162041.1 | 6 - 7 | 924 | 75 | ||
NM_001168571.1 | NP_001162043.1 | 6 - 7 | 999 | 75 | ||
NM_018737.5 | NP_061207.1 | 6 - 7 | 999 | 75 | ||
NR_033143.1 | 6 - 7 | 889 | 75 | |||
GenBank mRNA | AK032018.1 | - | 6 - 7 | 726 | 75 | |
AK052854.1 | - | 6 - 7 | 915 | 75 | ||
AK084595.1 | - | 6 - 7 | 796 | 75 | ||
AK085381.1 | - | 6 - 7 | 774 | 75 | ||
AK150966.1 | - | 6 - 7 | 866 | 75 | ||
AK151597.1 | - | 6 - 7 | 866 | 75 | ||
AK161564.1 | - | 6 - 7 | 837 | 75 | ||
AK164206.1 | - | 6 - 7 | 826 | 75 | ||
BC003257.1 | - | 6 - 7 | 707 | 3500778 | 75 | |
U49385.1 | - | 6 - 7 | 889 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |